Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGATGCCGTCAGAATTGTTCAC[A/G]TTCATTCTGTCATCATCTTACGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300617 MIM: 300116 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRCC3 - BRCA1/BRCA2-containing complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018055.2 | 341 | Intron | NP_001018065.1 | |||
NM_001242640.1 | 341 | Intron | NP_001229569.1 | |||
NM_024332.3 | 341 | Intron | NP_077308.1 | |||
XM_005274751.4 | 341 | Missense Mutation | ATT,GTT | I,V 75 | XP_005274808.1 | |
XM_017029838.1 | 341 | Intron | XP_016885327.1 |
CMC4 - C-X9-C motif containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTCP1 - mature T-cell proliferation 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |