Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGACGGGCGCGCGCGGCCCCTGCG[A/C]ACTCGGCCGCCGCCTGCGCGCGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159557 | ||||||||||||||||||||
Literature Links: |
AFF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AFF1 - AF4/FMR2 family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166693.2 | 26 | UTR 5 | NP_001160165.1 | |||
NM_001313959.1 | 26 | Intron | NP_001300888.1 | |||
NM_001313960.1 | 26 | Intron | NP_001300889.1 | |||
NM_005935.3 | 26 | Intron | NP_005926.1 | |||
XM_005263007.3 | 26 | Intron | XP_005263064.1 | |||
XM_005263009.3 | 26 | UTR 5 | XP_005263066.1 | |||
XM_005263011.3 | 26 | Intron | XP_005263068.1 | |||
XM_005263013.3 | 26 | UTR 5 | XP_005263070.1 | |||
XM_011531973.2 | 26 | UTR 5 | XP_011530275.1 | |||
XM_017008215.1 | 26 | UTR 5 | XP_016863704.1 | |||
XM_017008216.1 | 26 | Intron | XP_016863705.1 |
LOC100506746 - uncharacterized LOC100506746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |