Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCTCCAGCCCTAAAATGTAGCA[A/G]GTTATTGCTAGTATTTCTTTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610683 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBS12 - Bardet-Biedl syndrome 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178007.1 | Intron | NP_001171478.1 | ||||
NM_152618.2 | Intron | NP_689831.2 | ||||
XM_011531680.2 | Intron | XP_011529982.1 | ||||
XM_017007831.1 | Intron | XP_016863320.1 |
CETN4P - centrin 4, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |