Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCGCGCCCTGGAGCTGCTAGAG[A/G]TCCGGAAGCACAGCCCCGAGGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 139396 | ||||||||||||||||||||
Literature Links: |
GUCY1A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUCY1A3 - guanylate cyclase 1 soluble subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000856.5 | 261 | UTR 5 | NP_000847.2 | |||
NM_001130682.2 | 261 | UTR 5 | NP_001124154.1 | |||
NM_001130683.3 | 261 | UTR 5 | NP_001124155.1 | |||
NM_001130684.2 | 261 | Intron | NP_001124156.1 | |||
NM_001130685.2 | 261 | UTR 5 | NP_001124157.1 | |||
NM_001130687.2 | 261 | UTR 5 | NP_001124159.1 | |||
NM_001256449.1 | 261 | UTR 5 | NP_001243378.1 | |||
XM_005262955.2 | 261 | UTR 5 | XP_005263012.1 | |||
XM_005262956.2 | 261 | UTR 5 | XP_005263013.1 | |||
XM_005262957.2 | 261 | UTR 5 | XP_005263014.1 | |||
XM_006714196.2 | 261 | UTR 5 | XP_006714259.1 | |||
XM_006714197.2 | 261 | UTR 5 | XP_006714260.1 | |||
XM_006714198.2 | 261 | UTR 5 | XP_006714261.1 | |||
XM_011531900.2 | 261 | UTR 5 | XP_011530202.1 |