Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTACCTACTGAATTTTAAGAA[A/G]CGTGTGTTTCTTGCTGTATTCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAPGEF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAPGEF2 - Rap guanine nucleotide exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014247.2 | Intron | NP_055062.1 | ||||
XM_005263358.3 | Intron | XP_005263415.1 | ||||
XM_005263359.3 | Intron | XP_005263416.1 | ||||
XM_005263360.3 | Intron | XP_005263417.1 | ||||
XM_005263361.3 | Intron | XP_005263418.1 | ||||
XM_006714420.3 | Intron | XP_006714483.1 | ||||
XM_006714421.3 | Intron | XP_006714484.1 | ||||
XM_006714422.3 | Intron | XP_006714485.1 | ||||
XM_011532425.2 | Intron | XP_011530727.1 | ||||
XM_011532426.2 | Intron | XP_011530728.1 | ||||
XM_011532427.2 | Intron | XP_011530729.1 | ||||
XM_017008856.1 | Intron | XP_016864345.1 | ||||
XM_017008857.1 | Intron | XP_016864346.1 | ||||
XM_017008858.1 | Intron | XP_016864347.1 | ||||
XM_017008859.1 | Intron | XP_016864348.1 |