Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAATTGAAGACATTTACCACCCT[A/G]AGTAGGTCCTGCTGCTTCATTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD55 - CD55 molecule (Cromer blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000574.4 | Intron | NP_000565.1 | ||||
NM_001114752.2 | Intron | NP_001108224.1 | ||||
NM_001300902.1 | Intron | NP_001287831.1 | ||||
NM_001300903.1 | Intron | NP_001287832.1 | ||||
NM_001300904.1 | Intron | NP_001287833.1 | ||||
XM_017000467.1 | Intron | XP_016855956.1 |