Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATGCTCTAAGAAAATTTATAA[A/C]ATTAAGGCTGAAATTACCATGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607502 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DISP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DISP1 - dispatched RND transporter family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032890.3 | Intron | NP_116279.2 | ||||
XM_005273335.2 | Intron | XP_005273392.1 | ||||
XM_006711592.2 | Intron | XP_006711655.1 | ||||
XM_006711594.3 | Intron | XP_006711657.1 | ||||
XM_011510072.2 | Intron | XP_011508374.1 | ||||
XM_011510073.2 | Intron | XP_011508375.1 | ||||
XM_011510074.2 | Intron | XP_011508376.1 | ||||
XM_011510075.2 | Intron | XP_011508377.1 | ||||
XM_011510077.2 | Intron | XP_011508379.1 | ||||
XM_017002611.1 | Intron | XP_016858100.1 | ||||
XM_017002612.1 | Intron | XP_016858101.1 | ||||
XM_017002613.1 | Intron | XP_016858102.1 |