Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCAGTCATTTAGTTCTACTTGC[A/C]GTACTTCAAGAGGATCTTCACTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102595 | ||||||||||||||||||||
Literature Links: |
ACYP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACYP2 - acylphosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320586.1 | 118 | Intron | NP_001307515.1 | |||
NM_001320587.1 | 118 | Intron | NP_001307516.1 | |||
NM_001320588.1 | 118 | UTR 5 | NP_001307517.1 | |||
NM_001320589.1 | 118 | Intron | NP_001307518.1 | |||
NM_001320590.1 | 118 | Intron | NP_001307519.1 | |||
NM_138448.3 | 118 | Intron | NP_612457.1 | |||
XM_017005406.1 | 118 | Intron | XP_016860895.1 | |||
XM_017005407.1 | 118 | Intron | XP_016860896.1 | |||
XM_017005408.1 | 118 | Intron | XP_016860897.1 | |||
XM_017005409.1 | 118 | Intron | XP_016860898.1 | |||
XM_017005410.1 | 118 | Intron | XP_016860899.1 | |||
XM_017005411.1 | 118 | Intron | XP_016860900.1 | |||
XM_017005412.1 | 118 | Intron | XP_016860901.1 | |||
XM_017005413.1 | 118 | Intron | XP_016860902.1 |