Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAATTATAATGATGTCTACAGC[A/G]CCTTCCACGTCCTCTGTAAAGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASL11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASL11B - RAS like family 11 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023940.2 | 757 | Missense Mutation | ACC,GCC | T,A 193 | NP_076429.1 |
SCFD2 - sec1 family domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |