Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTTAAAGCAAAAAGCTCTGTT[C/T]CTGCCTCAGATGATGCCTATCCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604147 MIM: 605974 | ||||||||||||||||||||
Literature Links: |
PTTG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTTG1 - pituitary tumor-transforming 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282382.1 | 716 | Missense Mutation | CCT,TCT | P,S 105 | NP_001269311.1 | |
NM_001282383.1 | 716 | Missense Mutation | CCT,TCT | P,S 105 | NP_001269312.1 | |
NM_004219.3 | 716 | Missense Mutation | CCT,TCT | P,S 105 | NP_004210.1 |
SLU7 - SLU7 homolog, splicing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |