Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACATACTGAACTAGTGTTTAA[C/T]GGCTCTGAGAATAAAACCAAACCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610076 | ||||||||||||||||||||
Literature Links: |
CDK20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK20 - cyclin dependent kinase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039803.2 | 1793 | UTR 3 | NP_001034892.1 | |||
NM_001170639.1 | 1793 | UTR 3 | NP_001164110.1 | |||
NM_001170640.1 | 1793 | UTR 3 | NP_001164111.1 | |||
NM_012119.4 | 1793 | UTR 3 | NP_036251.2 | |||
NM_178432.3 | 1793 | UTR 3 | NP_848519.1 | |||
XM_017014561.1 | 1793 | UTR 3 | XP_016870050.1 | |||
XM_017014562.1 | 1793 | UTR 3 | XP_016870051.1 | |||
XM_017014563.1 | 1793 | UTR 3 | XP_016870052.1 | |||
XM_017014564.1 | 1793 | UTR 3 | XP_016870053.1 |