Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGTAGATGATCCATGTTTAGA[A/T]CGGTCCATTATCTATAAATTATACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611128 | ||||||||||||||||||||
Literature Links: |
MDGA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MDGA2 - MAM domain containing glycosylphosphatidylinositol anchor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113498.2 | Intron | NP_001106970.3 | ||||
NM_182830.4 | Intron | NP_878250.2 | ||||
XM_011536519.2 | Intron | XP_011534821.1 | ||||
XM_011536522.2 | Intron | XP_011534824.1 | ||||
XM_011536525.2 | Intron | XP_011534827.1 | ||||
XM_017021060.1 | Intron | XP_016876549.1 | ||||
XM_017021061.1 | Intron | XP_016876550.1 |