Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTCCCTTTCCTTATGGAAATG[C/T]AAGAATAAAATATTTCATTAAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613279 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EML5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EML5 - echinoderm microtubule associated protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183387.2 | 7078 | UTR 3 | NP_899243.1 | |||
XM_006720070.3 | 7078 | UTR 3 | XP_006720133.1 | |||
XM_011536528.2 | 7078 | UTR 3 | XP_011534830.1 | |||
XM_011536530.2 | 7078 | UTR 3 | XP_011534832.1 | |||
XM_011536531.2 | 7078 | UTR 3 | XP_011534833.1 | |||
XM_011536532.2 | 7078 | UTR 3 | XP_011534834.1 | |||
XM_011536533.2 | 7078 | UTR 3 | XP_011534835.1 | |||
XM_011536534.2 | 7078 | UTR 3 | XP_011534836.1 | |||
XM_011536536.2 | 7078 | Intron | XP_011534838.1 | |||
XM_011536538.2 | 7078 | Intron | XP_011534840.1 | |||
XM_017021062.1 | 7078 | UTR 3 | XP_016876551.1 | |||
XM_017021063.1 | 7078 | UTR 3 | XP_016876552.1 | |||
XM_017021064.1 | 7078 | UTR 3 | XP_016876553.1 | |||
XM_017021065.1 | 7078 | UTR 3 | XP_016876554.1 | |||
XM_017021066.1 | 7078 | UTR 3 | XP_016876555.1 | |||
XM_017021067.1 | 7078 | UTR 3 | XP_016876556.1 | |||
XM_017021068.1 | 7078 | UTR 3 | XP_016876557.1 | |||
XM_017021069.1 | 7078 | UTR 3 | XP_016876558.1 | |||
XM_017021070.1 | 7078 | Intron | XP_016876559.1 |
Set Membership: |
HapMap |