Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGCTCAGTCTTGGCACTGACGC[C/T]GGAGGCCATCTCCAGCTCCCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605799 MIM: 614062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMN - amnion associated transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030943.3 | 5414 | Intron | NP_112205.2 |
CDC42BPB - CDC42 binding protein kinase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006035.3 | 5414 | UTR 3 | NP_006026.3 | |||
XM_005268227.1 | 5414 | UTR 3 | XP_005268284.1 | |||
XM_005268228.1 | 5414 | UTR 3 | XP_005268285.1 | |||
XM_005268229.1 | 5414 | UTR 3 | XP_005268286.1 | |||
XM_005268230.4 | 5414 | Intron | XP_005268287.1 | |||
XM_011537387.1 | 5414 | Intron | XP_011535689.1 |