Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCGGCCATGCCACACATCTGTG[C/T]TTGCCTGGCTGTTTCTGCACCGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP170B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP170B - centrosomal protein 170B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112726.2 | Intron | NP_001106197.1 | ||||
NM_015005.2 | Intron | NP_055820.2 | ||||
XM_005267550.3 | Intron | XP_005267607.1 | ||||
XM_011536665.2 | Intron | XP_011534967.1 | ||||
XM_011536666.2 | Intron | XP_011534968.1 | ||||
XM_011536667.2 | Intron | XP_011534969.1 | ||||
XM_017021227.1 | Intron | XP_016876716.1 | ||||
XM_017021228.1 | Intron | XP_016876717.1 | ||||
XM_017021229.1 | Intron | XP_016876718.1 | ||||
XM_017021230.1 | Intron | XP_016876719.1 |