Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCGAGCGCTGCGGGAAGACACT[G/T]ACCCCCGGCGGGCACGCGGAGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123875 MIM: 601183 MIM: 603526 | ||||||||||||||||||||
Literature Links: |
CRIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRIP1 - cysteine rich protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRIP2 - cysteine rich protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270837.1 | 1012 | Silent Mutation | CTG,CTT | L,L 234 | NP_001257766.1 | |
NM_001270841.1 | 1012 | Silent Mutation | CTG,CTT | L,L 39 | NP_001257770.1 | |
NM_001312.3 | 1012 | Silent Mutation | CTG,CTT | L,L 160 | NP_001303.1 |
MTA1 - metastasis associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |