Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGCTCAGGCGGGGATGTGTGCG[A/G]AGCCTGCCGCTGCTGCAGCGAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182279 | ||||||||||||||||||||
Literature Links: |
SNRPN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNRPN - small nuclear ribonucleoprotein polypeptide N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003097.4 | 66 | UTR 5 | NP_003088.1 | |||
NM_022805.3 | 66 | Intron | NP_073716.1 | |||
NM_022806.3 | 66 | Intron | NP_073717.1 | |||
NM_022807.3 | 66 | Intron | NP_073718.1 | |||
NM_022808.3 | 66 | Intron | NP_073719.1 |
SNURF - SNRPN upstream reading frame | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005678.4 | 66 | UTR 5 | NP_005669.2 | |||
NM_022804.2 | 66 | UTR 5 | NP_073715.1 |