Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGTGAGCCAGGATCCCCAGCTC[C/T]GCCTGTGGCCCCCCGGCCCCGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600447 MIM: 601210 | ||||||||||||||||||||
Literature Links: |
MAP3K12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K12 - mitogen-activated protein kinase kinase kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193511.1 | 2180 | Missense Mutation | AGA,GGA | R,G 661 | NP_001180440.1 | |
NM_006301.3 | 2180 | Missense Mutation | AGA,GGA | R,G 628 | NP_006292.3 | |
XM_005269138.3 | 2180 | Missense Mutation | AGA,GGA | R,G 661 | XP_005269195.1 | |
XM_005269140.2 | 2180 | Intron | XP_005269197.1 | |||
XM_006719588.3 | 2180 | Missense Mutation | AGA,GGA | R,G 661 | XP_006719651.1 | |
XM_011538725.2 | 2180 | Missense Mutation | AGA,GGA | R,G 661 | XP_011537027.1 | |
XM_017019956.1 | 2180 | Missense Mutation | AGA,GGA | R,G 628 | XP_016875445.1 |
PCBP2 - poly(rC) binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |