Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAATTCCGAAGCAATCTTCCTG[C/T]TAAGGCCATTGGACACAGAATCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612477 MIM: 609380 MIM: 180468 | ||||||||||||||||||||
Literature Links: |
IQCG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IQCG - IQ motif containing G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134435.2 | 354 | Intron | NP_001127907.1 | |||
NM_001323027.1 | 354 | Intron | NP_001309956.1 | |||
NM_001323028.1 | 354 | Intron | NP_001309957.1 | |||
NM_001323029.1 | 354 | Intron | NP_001309958.1 | |||
NM_001323030.1 | 354 | Intron | NP_001309959.1 | |||
NM_032263.4 | 354 | Intron | NP_115639.1 | |||
XM_017007316.1 | 354 | Intron | XP_016862805.1 | |||
XM_017007317.1 | 354 | Intron | XP_016862806.1 |
LMLN - leishmanolysin like peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL35A - ribosomal protein L35a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000996.2 | 354 | Missense Mutation | NP_000987.2 | |||
NM_001316311.1 | 354 | Intron | NP_001303240.1 |