Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACACTCACTGGATCTGCCATTC[A/T]AGATATGCGCAGTTCAGCACCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611806 MIM: 616600 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AS3MT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AS3MT - arsenite methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020682.3 | Intron | NP_065733.2 |
BORCS7 - BLOC-1 related complex subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BORCS7-ASMT - BORCS7-ASMT readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |