Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATGAAAAAATGCTTAAGCTTAT[A/G]CTTTCATTTGCCACTATTAAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153622 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSR1 - macrophage scavenger receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002445.3 | 2795 | Intron | NP_002436.1 | |||
NM_138715.2 | 2795 | UTR 3 | NP_619729.1 | |||
NM_138716.2 | 2795 | UTR 3 | NP_619730.1 |