Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123691 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYZ - crystallin zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130042.1 | 1777 | UTR 3 | NP_001123514.1 | |||
NM_001130043.1 | 1777 | UTR 3 | NP_001123515.1 | |||
NM_001134759.1 | 1777 | UTR 3 | NP_001128231.1 | |||
NM_001889.3 | 1777 | UTR 3 | NP_001880.2 | |||
XM_005270491.4 | 1777 | UTR 3 | XP_005270548.1 | |||
XM_011540747.1 | 1777 | UTR 3 | XP_011539049.1 | |||
XM_017000367.1 | 1777 | UTR 3 | XP_016855856.1 | |||
XM_017000368.1 | 1777 | UTR 3 | XP_016855857.1 |