Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCGTCCGCGCCGCGAGGAGGAG[A/G]CCCTGCTGGTTTCTGTGCGGGTGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612409 MIM: 602571 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RBM14 - RNA binding motif protein 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM14-RBM4 - RBM14-RBM4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198845.1 | 116 | Intron | NP_001185774.1 | |||
NM_001198846.1 | 116 | Intron | NP_001185775.1 |
RBM4 - RNA binding motif protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198843.1 | 116 | UTR 5 | NP_001185772.1 | |||
NM_001198844.1 | 116 | UTR 5 | NP_001185773.1 | |||
NM_002896.3 | 116 | UTR 5 | NP_002887.2 |
Set Membership: |
HapMap |