Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGTGCTGCAGGGCCCACAGCT[A/G]CAATGGGCAGAGCTGGAAATTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NQO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NQO2 - NAD(P)H quinone dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000904.4 | Intron | NP_000895.2 | ||||
NM_001290221.1 | Intron | NP_001277150.1 | ||||
NM_001290222.1 | Intron | NP_001277151.1 | ||||
NM_001318940.1 | Intron | NP_001305869.1 |