Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGTGTAACACTGATTGTGAAG[A/G]CCTACAGCTTTGTTCCTTCAAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMG4 - proteasome assembly chaperone 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128591.1 | 320 | Intron | NP_001122063.1 | |||
NM_001128592.1 | 320 | Intron | NP_001122064.1 | |||
NM_001135750.1 | 320 | Intron | NP_001129222.1 | |||
XM_011514591.2 | 320 | UTR 5 | XP_011512893.1 | |||
XM_011514593.2 | 320 | Intron | XP_011512895.1 | |||
XM_011514594.2 | 320 | Intron | XP_011512896.1 | |||
XM_011514595.2 | 320 | UTR 5 | XP_011512897.1 |