Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATAGTTGCCCCCACCCTGGCTG[C/T]GATCCTGACGCACAGGCAGTTCTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609012 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR5 - WD repeat domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017588.2 | Intron | NP_060058.1 | ||||
NM_052821.3 | Intron | NP_438172.1 | ||||
XM_005272163.1 | Intron | XP_005272220.1 |