Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGTGCCTGTGCCTCCCTCACTTG[A/C]GCTGTGCCCCCCTCACCTGTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606801 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAEA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAEA - macrophage erythroblast attacher | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017405.2 | Intron | NP_001017405.1 | ||||
NM_001297430.1 | Intron | NP_001284359.1 | ||||
NM_001297431.1 | Intron | NP_001284360.1 | ||||
NM_001297432.1 | Intron | NP_001284361.1 | ||||
NM_001297433.1 | Intron | NP_001284362.1 | ||||
NM_005882.4 | Intron | NP_005873.2 | ||||
XM_006713849.3 | Intron | XP_006713912.1 | ||||
XM_006713850.2 | Intron | XP_006713913.1 | ||||
XM_011513376.1 | Intron | XP_011511678.1 | ||||
XM_011513377.1 | Intron | XP_011511679.1 | ||||
XM_017007636.1 | Intron | XP_016863125.1 | ||||
XM_017007637.1 | Intron | XP_016863126.1 |