Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTAAATAGCAAGAATATACAC[C/T]CTATTTCACAGTTTCTGTAGGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF595 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF595 - zinc finger protein 595 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286052.1 | Intron | NP_001272981.1 | ||||
NM_001286053.1 | Intron | NP_001272982.1 | ||||
NM_001286054.1 | Intron | NP_001272983.1 | ||||
NM_182524.3 | Intron | NP_872330.1 | ||||
XM_011513398.2 | Intron | XP_011511700.1 |