Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTTCGTGCCACTCCCGTCCCCA[C/T]CCCCACCCCAGTGTGTATAAGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300961 | ||||||||||||||||||||
Literature Links: |
MID1IP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MID1IP1 - MID1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098790.1 | 3452 | UTR 3 | NP_001092260.1 | |||
NM_001098791.1 | 3452 | UTR 3 | NP_001092261.1 | |||
NM_021242.5 | 3452 | UTR 3 | NP_067065.1 |
MID1IP1-AS1 - MID1IP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |