Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTTCTTCGGATTGATCAGCTA[C/A]TGGTGGAGTTAAAATTTCACCTGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600264 MIM: 616088 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AVPR1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AVPR1B - arginine vasopressin receptor 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf186 - chromosome 1 open reading frame 186 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007544.3 | Intron | NP_001007545.1 | ||||
XM_005272738.3 | Intron | XP_005272795.1 |