Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTAACGAGTAGGAAAGGTCAG[A/G]CAGGAACGGGGTGTGGGGAGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607779 MIM: 151442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAQR7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAQR7 - progestin and adipoQ receptor family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STMN1 - stathmin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145454.2 | Intron | NP_001138926.1 | ||||
NM_005563.3 | Intron | NP_005554.1 | ||||
NM_203399.1 | Intron | NP_981944.1 | ||||
NM_203401.1 | Intron | NP_981946.1 |