Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCACCACCTCGCGGTCGATCTC[A/T]GGATGGGTCCTGCCCTTGGTCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604057 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS3ST3A1 - heparan sulfate-glucosamine 3-sulfotransferase 3A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006042.2 | 1932 | Silent Mutation | CCA,CCT | P,P 373 | NP_006033.1 | |
XM_011524114.2 | 1932 | Silent Mutation | CCA,CCT | P,P 174 | XP_011522416.1 | |
XM_017025480.1 | 1932 | Intron | XP_016880969.1 |