Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCCCTTCTTCCGACAGCCGAT[G/T]CCTTTCATCACCACCATTCTGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGALS9 - galectin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002308.3 | Intron | NP_002299.2 | ||||
NM_009587.2 | Intron | NP_033665.1 | ||||
XM_006721892.2 | Intron | XP_006721955.1 | ||||
XM_006721893.2 | Intron | XP_006721956.1 | ||||
XM_006721894.3 | Intron | XP_006721957.1 | ||||
XM_006721895.3 | Intron | XP_006721958.1 | ||||
XM_011524796.1 | Intron | XP_011523098.1 | ||||
XM_017024623.1 | Intron | XP_016880112.1 |