Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCGCGCTCCCGGGACCATGTC[C/T]GCGCTGGCGGGTAAGGAAGGGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIMK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIMK2 - LIM domain kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031801.1 | 151 | Intron | NP_001026971.1 | |||
NM_005569.3 | 151 | Silent Mutation | TCC,TCT | S,S 2 | NP_005560.1 | |
NM_016733.2 | 151 | Intron | NP_057952.1 |
RNF185 - ring finger protein 185 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |