Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAGACAGATCATAAAAGGTGAG[A/T]GTGGTGCAGCTAATGACAACATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300663 MIM: 303631 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG4A - autophagy related 4A cysteine peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COL4A6 - collagen type IV alpha 6 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287758.1 | Intron | NP_001274687.1 | ||||
NM_001287759.1 | Intron | NP_001274688.1 | ||||
NM_001287760.1 | Intron | NP_001274689.1 | ||||
NM_001847.3 | Intron | NP_001838.2 | ||||
NM_033641.3 | Intron | NP_378667.1 | ||||
XM_006724617.3 | Intron | XP_006724680.1 | ||||
XM_011530852.2 | Intron | XP_011529154.1 | ||||
XM_011530853.2 | Intron | XP_011529155.1 | ||||
XM_011530854.2 | Intron | XP_011529156.1 |