Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTGCCAGGATTGTTGCTGTGG[A/C]ATATCGAGTACCACTAACTTTTAAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 150370 MIM: 180646 | |||||||||||||||||||||||
Literature Links: |
RPSA PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RPSA - ribosomal protein SA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304288.1 | Intron | NP_001291217.1 | ||||
NM_002295.5 | Intron | NP_002286.2 |
SNORA6 - small nucleolar RNA, H/ACA box 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA62 - small nucleolar RNA, H/ACA box 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |