Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAGCATCCAGGCCTTCGCCAGC[A/G]GCTCCACACTGCACGGCCTGGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602866 | ||||||||||||||||||||
Literature Links: |
ASIC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASIC1 - acid sensing ion channel subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001095.3 | 149 | Missense Mutation | AGC,GGC | S,G 24 | NP_001086.2 | |
NM_001256830.1 | 149 | Intron | NP_001243759.1 | |||
NM_020039.3 | 149 | Missense Mutation | AGC,GGC | S,G 24 | NP_064423.2 | |
XM_011538350.1 | 149 | Missense Mutation | AGC,GGC | S,G 24 | XP_011536652.1 | |
XM_011538351.2 | 149 | Missense Mutation | AGC,GGC | S,G 24 | XP_011536653.1 | |
XM_011538352.1 | 149 | Missense Mutation | AGC,GGC | S,G 24 | XP_011536654.1 |