Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCTGGAAACCCTTCCCACAGA[A/G]GTCGCAGCCGAAGGGCTTGAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600871 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFI1 - growth factor independent 1 transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127215.1 | 1566 | Missense Mutation | CTC,TTC | L,F 400 | NP_001120687.1 | |
NM_001127216.1 | 1566 | Missense Mutation | CTC,TTC | L,F 400 | NP_001120688.1 | |
NM_005263.3 | 1566 | Missense Mutation | CTC,TTC | L,F 400 | NP_005254.2 | |
XM_005270749.3 | 1566 | Missense Mutation | CTC,TTC | L,F 400 | XP_005270806.1 | |
XM_011541245.2 | 1566 | Missense Mutation | CTC,TTC | L,F 400 | XP_011539547.1 | |
XM_011541246.2 | 1566 | Missense Mutation | CTC,TTC | L,F 400 | XP_011539548.1 |