Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTAAGTTTCTGTTCCATGGCTTC[C/T]GTTGCCAAACCTGTGGCTACAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 311010 MIM: 313440 | ||||||||||||||||||||
Literature Links: |
ARAF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARAF - A-Raf proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256196.1 | 581 | Missense Mutation | CGT,TGT | R,C 124 | NP_001243125.1 | |
NM_001256197.1 | 581 | Missense Mutation | CGT,TGT | R,C 124 | NP_001243126.1 | |
NM_001654.4 | 581 | Missense Mutation | CGT,TGT | R,C 124 | NP_001645.1 | |
XM_006724529.2 | 581 | Missense Mutation | CGT,TGT | R,C 124 | XP_006724592.1 | |
XM_011543906.2 | 581 | Missense Mutation | CGT,TGT | R,C 124 | XP_011542208.1 | |
XM_011543908.2 | 581 | Missense Mutation | CGT,TGT | R,C 124 | XP_011542210.1 | |
XM_011543909.2 | 581 | Intron | XP_011542211.1 |
SYN1 - synapsin I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |