Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGGTACAGACGTGCAGCATGT[C/T]TGCCAGAGGAAACTTGGGGCCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLAMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLAMF1 - signaling lymphocytic activation molecule family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003037.3 | 1771 | UTR 3 | NP_003028.1 | |||
XM_005245456.3 | 1771 | UTR 3 | XP_005245513.1 | |||
XM_011509905.2 | 1771 | UTR 3 | XP_011508207.1 | |||
XM_017002129.1 | 1771 | UTR 3 | XP_016857618.1 | |||
XM_017002130.1 | 1771 | UTR 3 | XP_016857619.1 | |||
XM_017002131.1 | 1771 | UTR 3 | XP_016857620.1 |