Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGCATTTTAAGTGCTCAGTAA[A/G]TACTAGCAATTATTAATTGTTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609874 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITLN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITLN2 - intelectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080878.2 | Intron | NP_543154.1 | ||||
XM_011509210.1 | Intron | XP_011507512.1 | ||||
XM_011509211.1 | Intron | XP_011507513.1 |
LOC101928372 - uncharacterized LOC101928372 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |