Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGCTTGAGGATGGCGGAATCTTC[A/G]CAGTGATCTATACATTGGCCATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602672 MIM: 603702 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP210L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUP210L - nucleoporin 210 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159484.1 | 5466 | Silent Mutation | TGC,TGT | C,C 1646 | NP_001152956.1 | |
NM_207308.2 | 5466 | Silent Mutation | TGC,TGT | C,C 1798 | NP_997191.2 | |
XM_011510122.1 | 5466 | Silent Mutation | TGC,TGT | C,C 1754 | XP_011508424.1 | |
XM_011510123.1 | 5466 | Silent Mutation | TGC,TGT | C,C 1745 | XP_011508425.1 | |
XM_011510124.1 | 5466 | Missense Mutation | GCG,GTG | A,V 1730 | XP_011508426.1 | |
XM_017002788.1 | 5466 | Intron | XP_016858277.1 | |||
XM_017002789.1 | 5466 | Intron | XP_016858278.1 |
RAB13 - RAB13, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS27 - ribosomal protein S27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |