Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGGGCCCTCTGAGGGGGGCTC[C/T]TCAAGCCCCGTGAGGTTGTCCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUAK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUAK2 - NUAK family kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030952.2 | 1163 | Silent Mutation | NP_112214.2 | |||
XM_005245515.4 | 1163 | Missense Mutation | XP_005245572.1 |