Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACGTAATTTAGTAACATGCATCA[A/G]CTATCCTAGAACATCTACAGGGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602462 MIM: 604831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRMP1 - collapsin response mediator protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014809.2 | 2436 | UTR 3 | NP_001014809.1 | |||
NM_001288661.1 | 2436 | UTR 3 | NP_001275590.1 | |||
NM_001288662.1 | 2436 | UTR 3 | NP_001275591.1 | |||
NM_001313.4 | 2436 | UTR 3 | NP_001304.1 |
EVC - EvC ciliary complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306090.1 | 2436 | Intron | NP_001293019.1 | |||
NM_001306092.1 | 2436 | Intron | NP_001293021.1 | |||
NM_153717.2 | 2436 | Intron | NP_714928.1 | |||
XM_006713865.3 | 2436 | Intron | XP_006713928.1 | |||
XM_006713866.3 | 2436 | Intron | XP_006713929.1 | |||
XM_011513419.2 | 2436 | Intron | XP_011511721.1 | |||
XM_017007883.1 | 2436 | Intron | XP_016863372.1 |