Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTCTCCACTGTCTGAAAGATG[C/T]CAGAGACAGCTCCAAAGTCTATGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613976 | ||||||||||||||||||||
Literature Links: |
FANCE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FANCE - Fanconi anemia complementation group E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021922.2 | 687 | Silent Mutation | TGC,TGT | C,C 160 | NP_068741.1 | |
XM_005248885.2 | 687 | Silent Mutation | TGC,TGT | C,C 160 | XP_005248942.1 | |
XM_005248886.2 | 687 | Silent Mutation | TGC,TGT | C,C 160 | XP_005248943.1 | |
XM_005248887.2 | 687 | Silent Mutation | TGC,TGT | C,C 160 | XP_005248944.1 | |
XM_005248888.3 | 687 | Silent Mutation | TGC,TGT | C,C 160 | XP_005248945.1 | |
XM_011514343.2 | 687 | Silent Mutation | TGC,TGT | C,C 62 | XP_011512645.1 |