Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTTTTATTTTATAGTTATGGG[A/G]CCCATTCACATAAACTTTTTAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606534 | ||||||||||||||||||||
Literature Links: |
CYP3A43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP3A43 - cytochrome P450 family 3 subfamily A member 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278921.1 | 182 | Missense Mutation | ACC,GCC | T,A 27 | NP_001265850.1 | |
NM_022820.4 | 182 | Missense Mutation | ACC,GCC | T,A 27 | NP_073731.1 | |
NM_057095.2 | 182 | Missense Mutation | ACC,GCC | T,A 27 | NP_476436.1 | |
NM_057096.3 | 182 | Missense Mutation | ACC,GCC | T,A 27 | NP_476437.1 | |
XM_017012542.1 | 182 | Intron | XP_016868031.1 | |||
XM_017012543.1 | 182 | Intron | XP_016868032.1 | |||
XM_017012544.1 | 182 | Intron | XP_016868033.1 | |||
XM_017012545.1 | 182 | Intron | XP_016868034.1 |