Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGCAGGTGTTGCACCAGCGCC[G/T]TGGTGGAGGCTCCCTGAATGACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf16 - chromosome 9 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CIZ1 - CDKN1A interacting zinc finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131015.1 | 2160 | Silent Mutation | AGG,CGG | R,R 597 | NP_001124487.1 | |
NM_001131016.1 | 2160 | Silent Mutation | AGG,CGG | R,R 653 | NP_001124488.1 | |
NM_001131017.1 | 2160 | Silent Mutation | AGG,CGG | R,R 592 | NP_001124489.1 | |
NM_001131018.1 | 2160 | Silent Mutation | AGG,CGG | R,R 573 | NP_001124490.1 | |
NM_001257975.1 | 2160 | Silent Mutation | AGG,CGG | R,R 683 | NP_001244904.1 | |
NM_001257976.1 | 2160 | Silent Mutation | AGG,CGG | R,R 552 | NP_001244905.1 | |
NM_012127.2 | 2160 | Silent Mutation | AGG,CGG | R,R 653 | NP_036259.2 | |
XM_005251888.3 | 2160 | Silent Mutation | AGG,CGG | R,R 653 | XP_005251945.2 | |
XM_005251891.3 | 2160 | Silent Mutation | AGG,CGG | R,R 597 | XP_005251948.2 | |
XM_005251892.3 | 2160 | Silent Mutation | AGG,CGG | R,R 592 | XP_005251949.2 | |
XM_005251893.4 | 2160 | Silent Mutation | AGG,CGG | R,R 573 | XP_005251950.4 | |
XM_006717039.3 | 2160 | Silent Mutation | AGG,CGG | R,R 624 | XP_006717102.2 | |
XM_011518481.1 | 2160 | Silent Mutation | AGG,CGG | R,R 648 | XP_011516783.1 | |
XM_011518482.2 | 2160 | Silent Mutation | AGG,CGG | R,R 629 | XP_011516784.2 | |
XM_011518483.1 | 2160 | Silent Mutation | AGG,CGG | R,R 629 | XP_011516785.1 | |
XM_017014594.1 | 2160 | Silent Mutation | AGG,CGG | R,R 683 | XP_016870083.1 | |
XM_017014595.1 | 2160 | Silent Mutation | AGG,CGG | R,R 648 | XP_016870084.1 | |
XM_017014596.1 | 2160 | Silent Mutation | AGG,CGG | R,R 597 | XP_016870085.1 |