Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACAGCTTGCTATGGCATTGTTCA[A/G]GTACCCACTGGACCGTGGTTTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLLT10 - myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195626.1 | 529 | Silent Mutation | CAA,CAG | Q,Q 60 | NP_001182555.1 | |
NM_001195627.1 | 529 | Silent Mutation | CAA,CAG | Q,Q 60 | NP_001182556.1 | |
NM_001195628.1 | 529 | Silent Mutation | CAA,CAG | Q,Q 60 | NP_001182557.1 | |
NM_001195630.1 | 529 | Silent Mutation | CAA,CAG | Q,Q 60 | NP_001182559.1 | |
NM_001324296.1 | 529 | Silent Mutation | CAA,CAG | Q,Q 60 | NP_001311225.1 | |
NM_001324297.1 | 529 | UTR 5 | NP_001311226.1 | |||
NM_004641.3 | 529 | Silent Mutation | CAA,CAG | Q,Q 60 | NP_004632.1 |