Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTTTACCACATCTGAATGTCA[A/G]TGAGGGTTTGGCAGTTTAAGAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608483 MIM: 180203 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKTIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKTIP - AKT interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012398.2 | Intron | NP_001012398.1 | ||||
NM_001308325.1 | Intron | NP_001295254.1 | ||||
NM_022476.3 | Intron | NP_071921.1 | ||||
XM_005256095.4 | Intron | XP_005256152.1 | ||||
XM_005256096.4 | Intron | XP_005256153.1 | ||||
XM_005256097.4 | Intron | XP_005256154.1 | ||||
XM_005256098.4 | Intron | XP_005256155.1 | ||||
XM_017023564.1 | Intron | XP_016879053.1 | ||||
XM_017023565.1 | Intron | XP_016879054.1 | ||||
XM_017023566.1 | Intron | XP_016879055.1 |
RBL2 - RB transcriptional corepressor like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |