Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTACAGTATTACCTGGCCAGAG[A/G]TCTGGTGAGAGGAATTTATAGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFG3L1P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFG3L1P - AFG3 like matrix AAA peptidase subunit 1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CENPBD1 - CENPB DNA-binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |